Replacement Orifices (Stainless Steel & Siltek® Treated), Restek
Supplier: Restek
24233 22098 24219 24234 24237 24235 24249 24247 24248 24218 24245 24246 24236 22099
10863-176EA 242.07 CAD
10863-176 10863-134 10862-558 10862-658 10862-548 10862-828 10862-552 10862-660 10862-562 10862-644 10862-798 10862-546 10862-796 10862-560
Replacement Orifices (Stainless Steel & Siltek® Treated), Restek
Chromatography Supplies Chromatography Spare Parts
Use these orifices with a Veriflo® 423XL flow controller to change the flow range for alternative sampling times.
  • Range of standard gauge is –30" Hg to 60 psi
Ordering information: Do not exceed canister maximum pressure of 40 psig (2.75 bar).