Miniature Air Sampling Canisters (Electropolished Stainless Steel and Siltek Treated), Restek
Supplier: Restek
24188 24207 24189 24190 24205
10862-534EA 546.68 CAD
10862-534 10862-636 10862-642 10862-716 10862-838 10862-844 10862-850 10862-960 10862-962 10863-084 10863-174
Miniature Air Sampling Canisters (Electropolished Stainless Steel and Siltek Treated), Restek
Samplers Air Samplers
These small canisters are designed for controlled sampling, such as personal air sampling, as an alternative to tube and pump samplers.
  • For controlled sampling
  • Alternative to tube and pump samplers
  • Treated or non-treated versions available
  • 400 cc or 1,000 cc options
The 1,000 cc canister is suitable for sampling volatile organic compounds in air according to methods TO-14A, TO-15, IP-1A, ASTM 5466, OSHA PV 2120, and NJ DEP Low Level TO-15.
Ideal for indoor air, personal, emergency response, or soil gas sampling. Available with quick-connect fitting that is compatible with sampling and analysis instruments. Proven long life, leak-free performance, and effortless operation of the RAVE™ valve is also available.
Restek offers these products in stainless steel or with Siltek® treatment, for greatest inertness. We continue to offer passive coating technologies that are unmatched in the air sampling industry—try a Siltek®-treated canister to achieve the ultimate in analyte stability.
Accessories information: Sampling belt is Fully adjustable up to 50". Mini-Can stand securely holds 2-3/4" diameter cans. Personal sample inlet features a convenient clip and PTFE reducing ferrule allows direct connection from 1/16" tubing to 1/4" flow controller without another fitting.
Caution: Do not exceed canister maximum pressure of 40 psig (2.75 bar).