Vented Screw Caps, Non sterile, Chemglass
Supplier: Chemglass
CLS-1484-10
30623-042EA 16.67 CAD
30623-042
Vented Screw Caps, Non sterile, Chemglass
Closures Bottle Closures
Polypropylene vented cap has a 0.22 µm (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.
  • Autoclavable
  • Liner-less plug seal design for efficient sealing
  • GL-45 screw thread type