Test Tube Brushes, Justman
Supplier: Justman Brush
J610256 J610132 J710122 J710158 J610156 J710150 J610164
17140-006PK 289.12 CAD
17140-006 17145-002 17080-020 17145-005 17145-080 30620-538 30620-550
Test Tube Brushes, Justman
Brushes
Test tube brush is especially designed to clean the inner walls and bottom of test tubes to remove every trace of the chemical stored in it.
  • White-colored bristle with silver handle
  • Test tube brush with a radial tip
The tip of this brush features formed sprays that thoroughly clean the bottom of the test tube without damaging or scratching the glassware.