Antistatic Weigh Dishes, Simport Scientific
Supplier: Simport Scientific
D250-2 D250-1 D250-3
CA11006-614CS 67.5 CAD
CA11006-614 76299-236 76299-238
Antistatic Weigh Dishes, Simport Scientific
Boats Weighing Boats
Weighing Dishes will resist diluted acids, aqueous solutions, alcohols and bases, ideal for many applications such as weighing, dispensing, or storing.
  • Safe, contaminant-free, biologically inert economical containers
  • For weighing liquid or powdered samples in the laboratory
  • Flat bottom ensures perfect stability on countertops
Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Will withstand temperatures up to 80 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable