CellMaxx™ Bovine Serum Albumin (BSA), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219989710 0219989725 0219989750 0219989780 0219989790 0219989791
10273-266EA 240.32 CAD
10273-266 10273-268 10273-270 10273-272 10273-274 10273-276
CellMaxx™ Bovine Serum Albumin (BSA), MP Biomedicals
Proteins and Peptides
CellMaxx™ Bovine Albumin is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. This albumin has very low endotoxin and low IgG to simplify downstream purification, is tested for the absence of viruses according to 9CFR and has the highest possible biosafety level. The albumin supports growth of stem cells and increases protein production from hybridoma cells.
  • Mycoplasma, Virus: Not Detected
CellMaxx™ Bovine Albumin supports growth of stem cells and increases protein production from hybridoma cells. Albumin is also used to solubilize lipids, and as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present such as electrophoresis.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Bioburden: ≤100 cfu/g, IgG: ≤0.05 mg/g Total Protein, Origin: New Zealand; Moisture content: ≤5%; Isoelectric point: Endogenous material- 4.7, 4.9; Fatty acid depleted- 5.3 (in Water at 25 °C)(Lit.)