Bovine Serum Albumin (BSA), MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0218062010 0218062025 0218062091 0218062050 0218062080 0218062090
10252-990EA 209.41 CAD
10252-990 10252-992 10273-260 10273-320 10273-322 10273-324
Bovine Serum Albumin (BSA), MP Biomedicals
Proteins and Peptides

Microbiological Grade, =97% Total Protein,


  • Moisture: 1.60%


Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.


Bioburden: ≤100 cfu/g; Moisture content: ≤5%

Order Now

SPECIFICATIONS

Protein/peptide name Bovine Serum Albumin
Environmentally Preferable
Protein synonyms BSA|Fraction V|Cohn Fraction V
Species Bovine
Source Serum
CAS No. 9048-46-8
Endotoxin content ≤1 EU/mg
Bioburden <10 cfu/g
Purity 99%
Grade Microbiological Grade
Concentration 98%
Colour Yellow to Brown
Form Powder
Storage conditions 2-8 °C
Solubility 20% w/w in water
pH 6.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR