ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit, Genscript®
Supplier: Genscript
L00350 L00350C
95045-024EA 364.33 CAD
95045-024 10127-556
ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit, Genscript®
Microbiology Test Systems Endotoxin Tests
Genscript ToxinSensorTM Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.
  • Highly Sensitive detect endotoxin concentration in the range of 0.005 - 1 EU/mL
  • Incubation period can be shortened to 14 minutes
  • Color-stabilizer ensures accurate results
This method utilizes a modified Limulus Amebocyte Lysate and a synthetic color producing substrate to detect endotoxin chromogenically. In addition, any sample with color (e.g. cell bacterial culture medium, serum or blood) cannot be assayed by this kit.