VWR® Animal Ear Punches
Supplier: VWR International
11647-253 11647-254 11647-255 11647-256 11647-257
10806-286EA 42.13 CAD
10806-286 10806-288 10806-290 10806-292 10806-294
VWR® Animal Ear Punches
Biopsy Punches
VWR® Animal Ear Punches are made from corrosion-resistant ASTM (899-94) Series 400 stainless steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These ear punches are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.