ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Calbiochem®

Supplier: MilliporeSigma

481973-100MG 481973-1GM 481973-25MG 481973-500MG 481973-50MG
CA80053-342EA 410.06 CAD
CA80053-342 CA80053-344 CA80053-346 CA80053-348 CA80053-350
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form), Calbiochem®
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

A ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes.


It is generated by reduction of the electron acceptor NADP+. The following biological pathways involve NADPH: formation of carbohydrate from CO₂ during photosynthesis, maintenance of high levels of reduced glutathione in erythrocytes, reduction of thioredoxin.

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
Storage Temperature: Freezer
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR