PolarSafe™ Metal Freezer Boxes, Argos Technolgies
Supplier: Antyila Scientific
04398-10 04398-11 04398-08 04398-09
75800-320EA 51.53 CAD
75800-320 75800-322 75800-324 75800-326
PolarSafe™ Metal Freezer Boxes, Argos Technolgies
Boxes Cryoboxes
PolarSafe™ Metal Freezer Boxes are available in stainless steel or aluminum formats
Both the stainless steel and aluminum metal freezer boxes are guaranteed not to rust. They can be used in most liquid nitrogen tanks available on the market.
Drain holes in each box make them ideal for storing in liquid nitrogen vapor environments. Boxes are available in two sizes, with the option of 2" or 3" box heights. Metal boxes can be stored in the same rack configurations as 2" or 3" cardboard boxes. Cardboard dividers can be added depending on the application.