PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
73818A-24 73818-15 73818-18 73818-20 73818-24 73818X-18 N73818T-13 774161-0008 774161-0013 774161-0015 774161-0018 774161-0020 774161-0024 774161-0924 73818X-24
66010-751CS 410.43 CAD
66010-751 66010-774 66010-785 66010-796 66010-945 66010-946 93001-096 KT774161-0008 KT774161-0013 KT774161-0015 KT774161-0018 KT774161-0020 KT774161-0024 KT774161-0924 66010-947
PTFE-Faced Silicone Rubber Septa, Autoclavable, DWK Life Sciences
Septa
Ideal for use with open-top caps to access container contents with a syringe.
  • Resists coring
  • Highly chemically-resistant PTFE facing maintains integrity of content
  • Silicone rubber backing allows repeated puncturing through the seal
  • Recommended for use in all autosamplers using screw thread vials