Micro-organism Lysing VK01, Precellys® Lysing Kit, Bertin
Supplier: BERTIN CORP
P000914-LYSK0-A.0 P000929-LYSK0-A.0
10144-504EA 459.24 CAD
10144-504 10144-562
Micro-organism Lysing VK01, Precellys® Lysing Kit, Bertin
Beads Glass Beads
VK01 is made with 0.1 mm inert glass beads and designed for micro-organism lysing like gram-, gram+ bacteria, spores and more.
  • Ideal for yeast, bacteria and fungi lysis
  • Dnase/Rnase free
  • Autoclavable
Use with the Precellys®24, Precellys® Evolution, or Minilys homogenizers.
50 tubes (prefilled)
Certifications: Dnase/Rnase free certified.
Delivery information: Packaged 1 kit contain 50 prefilled and ready-to-use tubes.