Stericheck Closed Sterility Test System, Type ML, MDI
Supplier: MDI MEMBRANE TECHNOLOGIES, INC.
SMLV0902XXX302 SMLC0902XXX302 SMLV0902XXX202 SMLC0902XXX202
75796-902CS 3072.09 CAD
75796-902 75796-904 75798-778 75798-780
Stericheck Closed Sterility Test System, Type ML, MDI
Sterilizers Loop Sterilizers
Stericheck Type ML is a presterilized device for closed loop sterility testing of Medical Devices with Female Luer connections.
  • Raised vents prevent air flow obstruction due to fluid logging
  • Tough membrane helps withstand inadvertent back pressure
  • 'L' shaped flow directors prevent frothing
  • Archivable sterilization indicator in each pack
  • Validated for equal sample distribution
  • Can be used with the MDI Steripump or other available equipment
This male luer lock adaptor is designed for sterility testing of medical devices with female luer connectors