EXP® Hex-Head Fittings (EXP® Reusable Fittings for HPLC and UHPLC for 10-32 fittings and ¹/₁₆" tubing), Restek
Supplier: Restek
25927 25929 25931 25928 25930 25926
10803-384PK 1503.1 CAD
10803-384 10853-244 10853-260 10853-376 10853-400 10853-576
EXP® Hex-Head Fittings (EXP® Reusable Fittings for HPLC and UHPLC for 10-32 fittings and ¹/₁₆" tubing), Restek
Fittings and Connectors
Restek is pleased to offer the reusable EXP® fitting system from Optimize Technologies for the ultimate in easy, reliable LC connections.
  • Hand-tight fitting style achieves effortless HPLC seals—no tools needed for a 8,700+ psi seal
  • Both hand-tight and hex-head styles wrench-tighten for reliable UHPLC use up to 20,000+ psi
  • Patented ferrule can be installed repeatedly without compromising high-pressure seal
  • Hybrid design combines the durability of titanium with the sealing ability of PEEK
  • Cutting-edge system provides ZDV (zero dead volume) connection to any 1
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable