Waste Overflow Indicator for LC Systems, Restek
Supplier: Restek
26550 26543
10853-538EA 618.4 CAD
10853-538 10853-606
Waste Overflow Indicator for LC Systems, Restek
Closures Safety Caps and Safety Waste Caps
The Restek® waste overflow indicator will help keep your mobile phase waste where it belongs.
  • Avoid messy pooling around mobile phase waste containers
  • Audible alarm instantly alerts user, preventing overflow
  • Compact, battery-operated unit
  • Available for 4-liter and GL-45 solvent bottles
Compact, battery-operated unit fits securely on solvent bottles and accommodates two waste streams. An audible alarm is given as the solvent waste container approaches capacity, giving you time to empty or change the container. Another innovative design from Restek!
Ordering information: Bottle not included.