PEEK Fitting Extractor, Restek
Supplier: Restek
25325
RK25325EA 173.64 CAD
RK25325
PEEK Fitting Extractor, Restek
Puller and Inserter Tools
Drill into the broken fitting, then screw the extractor into the fitting and remove it easily.
Eliminates the need for heat or other techniques that could damage your column.