Column End-Fittings, Restek
Supplier: Restek
25077 25078
10853-344EA 83.4 CAD
10853-344 10853-622
Column End-Fittings, Restek
Fittings and Connectors
These 1/4" compression end-fittings are compatible with 1/16" connecting tubing using Valco® and Parker fittings.
  • Fittings with distribution cone are intended for use on 3.0–4.6 mm ID HPLC columns
  • Fittings with flat bottom are intended for use on 1.0–3.0 mm ID HPLC columns