Secure-Fit Fittings (Two Connectors), Restek
Supplier: Restek
25211 26513
10853-390EA 288.87 CAD
10853-390 10853-604
Secure-Fit Fittings (Two Connectors), Restek
Fittings and Connectors
A good connection between HPLC components is necessary for trouble-free chromatography.
Secure-Fit connectors from Restek and Selerity Technologies ensure a consistent, leak-free seal—and they eliminate excess dead volume! An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.