Secure-Fit Fittings (Single Connectors), Restek
Supplier: Restek
25236 25237 25182 25188 25181
10853-298EA 169.41 CAD
10853-298 10853-300 10853-490 10853-492 10853-494 10853-552
Secure-Fit Fittings (Single Connectors), Restek
Fittings and Connectors
A good connection between HPLC components is necessary for trouble-free chromatography.
Secure-Fit connectors from Restek and Selerity Technologies ensure a consistent, leak-free seal—and they eliminate excess dead volume! An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.