Membrane Microfiltration Glassware, Restek
Supplier: Restek
KT953761-0000 KT953771-0000 KT953826-0000 KT953781-0000 KT953825-0000 KT953751-0000 KT953753-0000 KT953828-0000 KT953829-0000 KT953827-0000 KT953830-0000 KT676001-4035
10853-248EA 184.28 CAD
10853-248 10853-250 10853-268 10853-290 10853-380 10853-406 10853-484 10853-540 10853-542 10853-580 10853-582 10853-674 10853-774
Membrane Microfiltration Glassware, Restek
Funnels Membrane Filter Funnels
This 47 mm filtration apparatus with fritted glass support is recommended for routine filtration of corrosive liquids and removal of particles from LC solvents.
The ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers. The support base has a coarse-porosity glass frit and an integral vacuum connection, located above the drip tip to prevent contamination of the vacuum line with filtrate droplets. Each apparatus includes a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base, and a filtration flask.