Backpressure Regulators, Restek
Supplier: Restek
25018 25020 25017
RK25018EA 618.03 CAD
RK25018 RK25020 RK25017
Backpressure Regulators, Restek
Gas Regulators
Backpressure regulators can improve detector performance by preventing bubble formation in the detector flow cell.
They also are useful in post-column reaction lines and between detectors and fraction collectors in preparatory work. Regulators are superior to more specific alternative solutions, like small-bore tubing, in which pressure varies with flow rate. Our end-of-line and flow-through backpressure regulators are adjustable to ensure constant backpressure over a wide range of mobile phase viscosities and flow rates. The end-of-line model is available with 1/4-28 plastic flange-type fittings or high-pressure 1/16" compression fittings; this design adjusts from 15 to 60 psi (103 to 414 kPa). The flow-through design has 1/16" compression fittings and is adjustable from 7 to 75 psi (48 to 517 kPa).