Mouse Recombinant IL-27/P28 (from E. coli)
Supplier: Fujifilm Irvine Scientific
200-50AF-100UG 200-50AF-10UG 200-50AF-1MG 200-50AF-500UG
10788-200EA 1497.78 CAD
10788-200 10788-202 10788-204 10788-206 10822-216
Mouse Recombinant IL-27/P28 (from E. coli)
Proteins and Peptides
The p28 subunit of interleukin 27 (IL-27), also known as interleukin 30 (IL-30), is a member of the interleukin 12 (IL-12) family of cytokines. p28 is a secreted polypeptide that associates with the Epstein-Barr virus induced gene 3 (EBI3) to form the IL-27 cytokine heterodimer complex. IL-27 functions as a proinflammatory cytokine that induces immunomodulatory effects in naïve CD4+ T cells, mast cells, and monocytes. p28 can also form a complex with cytokine-like factor 1 (CLF), that is secreted by dendritic cells, to regulate natural killer (NK) and T cell functions.
  • High quality
  • Low endotoxin
  • Affordable