Mouse Recombinant GDNF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-37AF-100UG 200-37AF-10UG 200-37AF-1MG 200-37AF-500UG
10788-114EA 1497.78 CAD
10788-114 10788-116 10788-118 10788-120 10822-016
Mouse Recombinant GDNF (from E. coli)
Proteins and Peptides
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
  • High quality
  • Low endotoxin
  • Affordable