Human Recombinant FGF-10 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-183AF-5UG 100-183AF-25UG 100-183AF-1MG 100-183AF-100UG 100-183AF-500UG
10788-592EA 151.36 CAD
10788-592 10788-594 10788-596 10788-598 10821-952 77660-696 77660-698
Human Recombinant FGF-10 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis. FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinizing epidermal cells, and induces the migration and invasion of cancer cells.
  • High quality
  • Low endotoxin
  • Affordable