Mouse Recombinant TPO (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Exodus-2, also known as CCL21 and 6Ckine, is a chemokine that is strongly produced in the human lymph nodes and spleen. Exodus-2 signals through the chemokine receptor CCR7 to regulate thymocyte and activated T cell migration. Exodus-2 also mediates the homing of lymphocytes to the lymphatic system. Human and mouse Exodus-2 proteins share greater than 85% amino acid sequence identity.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.