Mouse Recombinant IL-4 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-18AF-100UG
200-18AF-20UG
200-18AF-1MG
200-18AF-5UG
200-18AF-500UG
10789-144EA
980.06
CAD
10789-144
10789-146
10789-148
10789-150
10822-102
Mouse Recombinant IL-4 (from E. coli)
Proteins and Peptides
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
- High quality
- Low endotoxin
- Affordable