Mouse Recombinant GM-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-15AF-100UG
200-15AF-1MG
200-15AF-20UG
200-15AF-5UG
200-15AF-500UG
10787-942EA
893.9
CAD
10787-942
10787-944
10787-946
10787-948
10822-028
Mouse Recombinant GM-CSF (from E. coli)
Proteins and Peptides
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
- High quality
- Low endotoxin
- Affordable