Human Recombinant FLT-3 Ligand (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-21AF-10UG 100-21AF-1MG 100-21AF-100UG 100-21AF-500UG
10787-444EA 390.81 CAD
10787-444 10787-446 10787-448 10789-022 10821-978
Human Recombinant FLT-3 Ligand (from E. coli)
Proteins and Peptides
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signalling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7,) or in combination of IL-7 and interleukin 3 (IL-3). Human FLT-3 ligand is active on mouse cells.
  • Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.