Disposable Chromatography Syringe, Microsolv
Supplier: MicroSolv Technology Corp
58903-S-A 58903-S-C 58903-S-K 58905-S-A 58905-S-C 58905-S-K 58910-S-A 58910-S-C 58910-S-K 58920-S-A 58920-S-C 58920-S-K
97054-870PK 66.14 CAD
97054-870 97054-872 97054-874 97054-876 97054-878 97054-880 97054-882 97054-884 97054-886 97054-888 97054-890 97054-892
Disposable Chromatography Syringe, Microsolv
Syringes Syringes for Chromatography
These syringes are used in chromatography and dissolution testing
  • Safety stop to prevent accidental spills
  • For situation needing an inert, non-reactive syringe
Designed with centered, Luer Lock tips, these syringes simply are the best for laboratory applications such as HPLC, Dissolution Testing, CE, LCMS.
They are latex free, contain no rubber, no silicone oil, styrene or DEHP.
Order Now
SPECIFICATIONS
Environmentally Preferable