Human Recombinant PDGF-AA (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-16AF-100UG 100-16AF-10UG 100-16AF-1MG 100-16AF-500UG
10788-910EA 1095.48 CAD
10788-910 10788-912 10788-914 10788-916 10822-332
Human Recombinant PDGF-AA (Animal free) (from E. coli)
Proteins and Peptides
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.
  • High quality
  • Low endotoxin
  • Affordable
PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
Caution: For research use only.