Mouse Recombinant IL-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-01AF-100UG 200-01AF-10UG 200-01AF-1MG 200-01AF-500UG
10787-842EA 1260.62 CAD
10787-842 10789-064 10789-066 10789-068 10822-094
Mouse Recombinant IL-3 (from E. coli)
Proteins and Peptides
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
  • High quality
  • Low endotoxin
  • Affordable