Human Recombinant Omentin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-140AF-10UG 100-140AF-1MG 100-140AF-100UG
10787-298EA 1428.1 CAD
10787-298 10787-300 10787-302 10787-304 10822-324 77660-660
Human Recombinant Omentin (from E. coli)
Proteins and Peptides
Omentin is an adipokine that is produced and secreted by the small intestine, visceral adipose tissue, perivascular adipose tissue, and epicardial adipose tissue. Omentin enhances insulin-stimulated glucose uptake in adipocytes and is a link between obesity and Type 2 Diabetes. Omentin also functions as a vasodilator and plays a protective role during coronary atherosclerosis and hypertension.
  • Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.