Human Recombinant GM-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-08AF-100UG 100-08AF-1MG 100-08AF-20UG 100-08AF-5UG 100-08AF-500UG
10787-116EA 900.3 CAD
10787-116 10787-118 10787-120 10787-122 10822-024
Human Recombinant GM-CSF (from E. coli)
Proteins and Peptides
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
  • Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.