Human Recombinant M-CSF (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-03AF-100UG 100-03AF-10UG 100-03AF-1MG 100-03AF-500UG
10787-060EA 1158.12 CAD
10787-060 10787-062 10787-064 10787-066 10822-266
Human Recombinant M-CSF (Animal free) (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types.
  • High quality
  • Low endotoxin
  • Affordable
M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
Caution: For research use only.