Silicone oil, 100 - 300 mPa.s (20 °C) for oil baths, Supelco®

Supplier: MilliporeSigma

Synonyms: Polyphenyl-methylsiloxane

M-Clarity™ quality level = MQ100

1.07742.0100 1.07742.1000
CA10436-324EA 121 CAD
CA10436-324 CA10436-326
Silicone oil, 100 - 300 mPa.s (20 °C) for oil baths, Supelco®
Silicone oil

Silicone oil used as oil bath up to 250 °C.


  • Viscosity : 100 - 300 mPa.s (20 °C)


Packaging: Packed in plastic bottles.

Boiling Pt: >315 °C (1013 hPa)
Density: 1.04 g/cm³ (20 °C)
Flash Pt: >295 °C
MDL Number: MFCD00132673
CAS Number: 68083-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR