MES sodium salt ≥99%, ULTROL® biological buffer, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 2-morpholinoethane-1-sulfonate, Sodium 3-morpholin-1-ylethylsulphonate, Sodium 3-morpholin-1-ylethylsulfonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, Sodium 2-Morpholinoethanesulfonate, 2-(N-Morpholino)ethanesulfonic acid sodium salt, MES-Na

475894-100GM 475894-1KG
CA80058-422EA 357.56 CAD
CA80058-422 CA80058-424
MES sodium salt ≥99%, ULTROL® biological buffer, Calbiochem®
MES sodium salt

These Zwitterionic buffers are useful in the pH range of 5.5 to 6.7 and has a pKa of 6.10 at 25 °C.


Has an absorbance of ≤0.05 (100 mM, H₂O, 260 nM).

Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065473
CAS Number: 71119-23-8

Order Now

Specification Test Results

Purity ≥99% by titration (dry basis)
Heavy metals ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR