Protease Inhibitor Cocktail Set III, Animal-Free
Supplier: MilliporeSigma
535140-1SET
535140-1ML
CA80017-246EA
1318.76
CAD
CA80017-246
CA80017-248
Protease Inhibitor Cocktail Set III, Animal-Free
Cell Culture Supplements and Additives
A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine proteases as well as aminopeptidases.
- Storage temperature: −20 °C
This cocktail is recommended for use with mammalian cell and tissue extracts, but is also suitable for bacterial extracts. Supplied with a data sheet. Liquid form.