9000 Series Full Face Respirators, Moldex®
Supplier: Moldex/Metric
9001 9002 9003
10065-406BG 391.59 CAD
10065-406 10065-408 10065-410
9000 Series Full Face Respirators, Moldex®
Respirators Reusable Respirators Full Face Respirators
Full face respirators with technologically advanced features and exclusive over-molded lens design.
  • High-tech, streamlined design for exceptional worker acceptance
  • Exclusive over-molded lens offers greater field of vision
  • Lighter weight for superior all-day comfort
  • Fewer parts means less cost and easy maintenance
  • Stand-away head harness for easy donning and doffing
  • Lens stays away from flat surface when laid down to avoid scratching
The 9000 series full face respirators with technologically advanced features and exclusive over-molded lens design eliminates the usual heavy clamping frame to create an innovative full face with lighter weight, greater field of vision, fewer parts, minimal maintenance and completely free of metal parts.
Certifications: Lens meets ANSI Z87+ for high impact requirements.
Accessories information: The cartridges are NIOSH approved for ammonia and methylamine. They are 100% PVC free and used with gas proof googles and Moldex® 9000 face mass. Also used with N95 or P100 filters against particulates.
Ordering information: Tear away lens protector included.