SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Supplier: SP Industries
BP-1827-002
14203-552EA 665.12 CAD
14203-552
SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Flasks Filter Flasks
These microfiltration components are designed for the filtration of aqueous liquids
  • HPLC mobile phase filtration
  • Suitable for general laboratory microfiltration
The components are interchangeable with competitive product offerings and can be used with filtering manifolds.
They may be used for analysis of particulate or bacterial contamination.