Iron Standards for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70326.0100 1.70376.0100
CA1.70326.0100EA 146 CAD
CA1.70326.0100 CA1.70376.0100
Iron Standards for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Iron Standards
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
Order Now
SPECIFICATIONS
Environmentally Preferable
Hazard class description Corrosive Material