Chromium Standards for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70312.0100
1.70374.0100
CA1.70312.0100EA
146
CAD
CA1.70312.0100
CA1.70374.0100
Chromium Standards for ICP, Certipur®, MilliporeSigma
Standards
Cationic Standards
Chromium Standards
- Cr(NO₃)₃ in HNO₃
- Traceable to SRM from NIST
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.