Rudiger® Physiological Spine
Supplier: Rudiger
A207
470221-152EA 485.65 CAD
470221-152
Rudiger® Physiological Spine
Educational Materials Biology Educational Materials Human Biology Learning Activities
Lifelike accuracy cast from a natural spine.
  • Fully articulated and flexible
  • Soft discs compress like natural tissue
  • 5 year manufacturers warranty
Our spine is fully articulated and flexible to show vertebral movements and alignments. Comes with arteries, spinal cord, nerve ends and flexible femur heads. The sacrum is sectioned to show course of spinal nerves. Contains a softer disc material, that compresses like natural tissue, bulging outward when under pressure. With stand.