Vileda® Professional Sterile Roll-O-Matic® Sponge Refills, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
147546 147548
10746-444CS 570.76 CAD
10746-444 10746-466 10746-468
Vileda® Professional Sterile Roll-O-Matic® Sponge Refills, Vileda Professional-FHP
Mops
The new "Advanced Technology" foam is manufactured specifically for the controlled environment industry. This foam allows 25% more liquid absorption and two times more liquid release than competing foams. Each refill is completely autoclavable and resistant to gamma and ETO sterilization. Refills are individually packaged and lot number controlled. All Vileda® Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. Manufactured in the USA.
  • Original refills have been the standard in the professional and controlled
More Product Information