Terbium Standard for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
Certipur®
1.70358.0100 1.70358.0100
CA1.70358.0100CS 19164.75 CAD
CA1.70358.0100 EM1.70358.0100
Terbium Standard for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Terbium Standards
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.