Extran® AP 18 Liquid, Mild Alkaline Concentrate, Merck
Supplier: MilliporeSigma
1.40118.2500 1.40118.5000 1.40118.9010 1.40118.9025
EM1.40118.2500EA 167.48 CAD
EM1.40118.2500 EM1.40118.5000 EM1.40118.9010 EM1.40118.9025
Extran® AP 18 Liquid, Mild Alkaline Concentrate, Merck
Cleaning Agents Detergents and Surface Cleansers
Extran® AP 18 is a universal cleaning agent for the gentle cleaning of alkali-sensitive items. Suitable for cleaning items which cannot be tainted with allergenic detergents, e.g. jewellery, glasses. Extran® AP 18 mild alkaline does not foam even during heavy agitation in a washing machine.
  • Surfactant-free
  • Mild alkaline liquid
  • Chlorine-free
  • Phosphate-free
  • Free from odorants/dyestuffs
Extran® AP 18 mild alkaline is a liquid main cleaning agent with mild alkaline properties for automatic dosage.
Dosing
The ideal dosing depends on the hardness of theMore Product Information