Palladium 10% on activated carbon hydrogenation catalyst (oxidised form), for synthesis, unreduced, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07104.0005 8.07104.0010 8.07104.0010 8.07104.0005
CA8.07104.0005EA 594.79 CAD
EM8.07104.0010 CA8.07104.0005 EM8.07104.0005 CA8.07104.0010
Palladium 10% on activated carbon hydrogenation catalyst (oxidised form), for synthesis, unreduced, Sigma-Aldrich®
Palladium
Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR