Rhodium Standards for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
Certipur®
1.70345.0100 1.70345.0100
CA1.70345.0100EA 273.09 CAD
CA1.70345.0100 EM1.70345.0100
Rhodium Standards for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Rhodium Standards
Rh(NO₃)₃ in HNO₃
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.