Methyl toluene-4-sulfonate, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.08333.9025 8.08333.9025
EM8.08333.9025EA 2197.78 CAD
EM8.08333.9025 CA8.08333.9025
Methyl toluene-4-sulfonate, Sigma-Aldrich®
Methyl toluene-4-sulphonate
Formula: C₈H₁₀O₃S
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00008417
CAS Number: 80-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR