Sodium acetate ≥99.0% (by perchloric acid titration), Supelco®
Supplier: MilliporeSigma
Synonyms:
Acetic acid sodium salt
M-Clarity™ quality level = MQ300
1.06268.9025
EM1.06268.9025EA
8644.75
CAD
EM1.06268.9025
Sodium acetate ≥99.0% (by perchloric acid titration), Supelco®
Sodium acetate
Formula:
H₃CCOONa MW: 82.03 g/mol Boiling Pt: 881.4 °C (1013 hPa) Melting Pt: 324 °C Density: 1.52 g/cm3 (20 °C) Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...